siRNA Details
Gene Symbol: CNTLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017738.3 | NP_060208.2 | 18 | 3165 | ||
XM_005251492.1 | XP_005251549.1 | 18 | 3162 | |||
XM_006716793.3 | XP_006716856.1 | 18 | 2963 | |||
XM_011517941.2 | XP_011516243.1 | 18 | 3165 | |||
XM_017014839.1 | XP_016870328.1 | 18 | 3165 | |||
XM_017014840.1 | XP_016870329.1 | 18 | 3165 | |||
XM_017014841.1 | XP_016870330.1 | 17 | 2970 | |||
XM_017014842.1 | XP_016870331.1 | 18 | 3165 | |||
XM_017014843.1 | XP_016870332.1 | 18 | 3165 | |||
XM_017014844.1 | XP_016870333.1 | 11 | 1841 | |||
XM_017014845.1 | XP_016870334.1 | 9 | 2646 | |||
XM_017014846.1 | XP_016870335.1 | 6 | 1169 | |||
XM_017014847.1 | XP_016870336.1 | 6 | 1162 | |||
GenBank mRNA | AK000283.1 | - | 14 | 2260 | ||
AK021596.1 | - | 6 | 1174 | |||
AK308837.1 | - | 18 | 3117 | |||
BX647069.1 | - | 18 | 3124 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
