siRNA Details
Gene Symbol: CNTLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017738.3 | NP_060208.2 | 8 | 1288 | ||
XM_005251492.1 | XP_005251549.1 | 8 | 1288 | |||
XM_006716793.3 | XP_006716856.1 | 8 | 1086 | |||
XM_011517941.2 | XP_011516243.1 | 8 | 1288 | |||
XM_017014839.1 | XP_016870328.1 | 8 | 1288 | |||
XM_017014840.1 | XP_016870329.1 | 8 | 1288 | |||
XM_017014842.1 | XP_016870331.1 | 8 | 1288 | |||
XM_017014843.1 | XP_016870332.1 | 8 | 1288 | |||
GenBank mRNA | AK000283.1 | - | 4 | 383 | ||
AK308837.1 | - | 8 | 1243 | |||
BX647069.1 | - | 8 | 1246 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
