siRNA Details
Gene Symbol: C1orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017860.3 | NP_060330.2 | 1 | 249 | ||
GenBank mRNA | AK000526.1 | - | 1 | 258 | ||
AK301712.1 | - | 1 | 321 | |||
AK315680.1 | - | 1 | 261 | |||
AY358436.1 | - | 1 | 292 | |||
BC002469.2 | - | 1 | 249 | 3347975 |
