siRNA Details
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017875.2 | NP_060345.2 | 5 | 975 | ||
XM_006713214.1 | XP_006713277.1 | 6 | 938 | |||
XM_011533869.2 | XP_011532171.1 | 6 | 1838 | |||
XM_017006710.1 | XP_016862199.1 | 5 | 974 | |||
GenBank mRNA | AK000558.1 | - | 5 | 951 | ||
AK225388.1 | - | 5 | 927 | |||
BC013194.1 | - | 5 | 636 | 3918744 | ||
CR457242.1 | - | 5 | 574 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
