siRNA Details
Gene Symbol: NMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017881.2 | NP_060351.1 | 6 | 607 | ||
NR_023352.1 | 7 | 864 | ||||
XM_006717162.1 | XP_006717225.1 | 7 | 892 | |||
XM_006717163.2 | XP_006717226.1 | 7 | 927 | |||
XM_011518817.1 | XP_011517119.1 | 6 | 717 | |||
XM_011518818.1 | XP_011517120.1 | 6 | 1239 | |||
XM_017014868.1 | XP_016870357.1 | 8 | 1213 | |||
XM_017014869.1 | XP_016870358.1 | 7 | 1520 | |||
XM_017014870.1 | XP_016870359.1 | 6 | 1263 | |||
GenBank mRNA | AK000566.1 | - | 6 | 572 | ||
AK023413.1 | - | 5 | 680 | |||
AK097144.1 | - | 7 | 837 | |||
AY611480.1 | - | 5 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
