siRNA Details
Gene Symbol: NMRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017881.2 | NP_060351.1 | 6 | 578 | ||
NR_023352.1 | 7 | 835 | ||||
XM_006717162.1 | XP_006717225.1 | 7 | 863 | |||
XM_006717163.2 | XP_006717226.1 | 7 | 898 | |||
XM_011518817.1 | XP_011517119.1 | 6 | 688 | |||
XM_011518818.1 | XP_011517120.1 | 6 | 1210 | |||
XM_017014868.1 | XP_016870357.1 | 8 | 1184 | |||
XM_017014869.1 | XP_016870358.1 | 7 | 1491 | |||
XM_017014870.1 | XP_016870359.1 | 6 | 1234 | |||
GenBank mRNA | AK000566.1 | - | 6 | 543 | ||
AK023413.1 | - | 5 | 651 | |||
AK097144.1 | - | 7 | 808 | |||
AY611480.1 | - | 5 | 333 | |||
BC001366.2 | - | 6 | 486 | 3051165 | ||
CB135264.1 | - | 6 | 578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
