siRNA Details
Gene Symbol: C1orf123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304759.1 | NP_001291688.1 | 5 | 299 | ||
NM_001304760.1 | NP_001291689.1 | 4 | 215 | |||
NM_017887.2 | NP_060357.1 | 6 | 356 | |||
GenBank mRNA | AK000587.1 | - | 6 | 334 | ||
AK092734.1 | - | 4 | 1200 | |||
AK094371.1 | - | 1 | 1532 | |||
BC010908.1 | - | 6 | 338 | 4093763 | ||
BE889129.1 | - | 3 | 168 | |||
BI603716.1 | - | 5 | 292 | |||
CR457246.1 | - | 5 | 292 |
