siRNA Details
Gene Symbol: TMCO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017905.4 | NP_060375.4 | 8, 9 | 1712 | ||
XM_006719969.1 | XP_006720032.1 | 8, 9 | 1493 | |||
XM_011537498.2 | XP_011535800.1 | 8, 9 | 1836 | |||
XM_011537502.2 | XP_011535804.1 | 8, 9 | 1712 | |||
XM_017020629.1 | XP_016876118.1 | 8, 9 | 1712 | |||
XM_017020631.1 | XP_016876120.1 | 7, 8 | 1520 | |||
XM_017020632.1 | XP_016876121.1 | 7, 8 | 1520 | |||
GenBank mRNA | AK000630.1 | - | 8, 9 | 1609 | ||
AK074415.1 | - | 5, 6 | 738 | |||
AY358768.1 | - | 8, 9 | 1490 | |||
BC068515.1 | - | 8, 9 | 1710 | 4819377 |
