siRNA Details
Gene Symbol: MCUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017918.4 | NP_060388.2 | 6, 7 | 963 | ||
XM_006714246.3 | XP_006714309.1 | 6, 7 | 1074 | |||
GenBank mRNA | AK000654.1 | - | 4, 5 | 650 | ||
AK095936.1 | - | 6, 7 | 870 | |||
AK291098.1 | - | 6, 7 | 957 | |||
BC002633.2 | - | 6, 7 | 901 | 3613298 | ||
BM830891.1 | - | 2, 3 | 337 | |||
CR457234.1 | - | 4, 5 | 560 |
