siRNA Details
Gene Symbol: MCUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017918.4 | NP_060388.2 | 7 | 1045 | ||
XM_006714246.3 | XP_006714309.1 | 7 | 1156 | |||
GenBank mRNA | AK000654.1 | - | 5 | 732 | ||
AK095936.1 | - | 7 | 952 | |||
AK291098.1 | - | 7 | 1039 | |||
AL832311.1 | - | 2 | 916 | |||
BC002633.2 | - | 7 | 983 | 3613298 | ||
BM830891.1 | - | 3 | 419 | |||
CR457234.1 | - | 5 | 642 |
