siRNA Details
Gene Symbol: MCUB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017918.4 | NP_060388.2 | 4 | 511 | ||
XM_006714246.3 | XP_006714309.1 | 4 | 622 | |||
GenBank mRNA | AK000654.1 | - | 2 | 198 | ||
AK095936.1 | - | 4 | 418 | |||
AK291098.1 | - | 4 | 505 | |||
AK311030.1 | - | 4 | 375 | |||
BC002633.2 | - | 4 | 449 | 3613298 | ||
CN427680.1 | - | 4 | 515 | |||
CR457234.1 | - | 2 | 108 |
