siRNA Details
Gene Symbol: PID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017933.4 | NP_060403.3 | 2, 3 | 537 | ||
XM_017004404.1 | XP_016859893.1 | 2, 3 | 5468 | |||
GenBank mRNA | AK125359.1 | - | 2, 3 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
