siRNA Details
Gene Symbol: TMEM255A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017938.3 | NP_060408.3 | 6 | 662 | ||
XM_017029619.1 | XP_016885108.1 | 6 | 682 | |||
GenBank mRNA | AF450484.1 | - | 6 | 592 | ||
AK289684.1 | - | 6 | 553 | |||
BC047054.1 | - | 6 | 559 | 5313196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
