siRNA Details
Gene Symbol: MOCOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000740.1 | - | 6 | 1192 | ||
AK222886.1 | - | 6 | 1181 | |||
AL834481.1 | - | 4 | 890 | |||
BC012079.1 | - | 6 | 1184 | 4550285 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
