siRNA Details
Gene Symbol: NRDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017970.3 | NP_060440.2 | 6 | 1525 | ||
XM_011536886.1 | XP_011535188.1 | 6 | 1323 | |||
XM_017021394.1 | XP_016876883.1 | 6 | 1323 | |||
XM_017021395.1 | XP_016876884.1 | 7 | 1474 | |||
GenBank mRNA | AK000870.1 | - | 6 | 1323 | ||
AK024113.1 | - | 1 | 66 | |||
AK294958.1 | - | 6 | 1329 | |||
BC098568.1 | - | 6 | 1303 | 30706812 | ||
BC118673.1 | - | 3 | 858 | 40028659 |
