siRNA Details
Gene Symbol: NRDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017970.3 | NP_060440.2 | 10 | 2235 | ||
XM_011536886.1 | XP_011535188.1 | 11 | 2063 | |||
XM_017021394.1 | XP_016876883.1 | 11 | 2063 | |||
XM_017021395.1 | XP_016876884.1 | 11 | 2184 | |||
XM_017021396.1 | XP_016876885.1 | 10 | 1778 | |||
GenBank mRNA | AK024113.1 | - | 5 | 776 | ||
AK294958.1 | - | 10 | 2039 | |||
BC098568.1 | - | 10 | 2013 | 30706812 | ||
BC118673.1 | - | 7 | 1568 | 40028659 |
