siRNA Details
Gene Symbol: AIM1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039775.3 | NP_001034864.2 | 2 | 1483 | ||
XM_005245918.2 | XP_005245975.1 | 2 | 1469 | |||
XM_011541672.1 | XP_011539974.1 | 2 | 1469 | |||
XM_011541673.2 | XP_011539975.1 | 2 | 1708 | |||
GenBank mRNA | AK000902.1 | - | 2 | 1482 | ||
DA456217.1 | - | 1 | 24 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
