siRNA Details
Gene Symbol: TAPBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002056.2 | - | 2 | 247 | ||
AK222956.1 | - | 2 | 247 | |||
AK223163.1 | - | 2 | 277 | |||
BC015017.2 | - | 2 | 286 | 3916213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
