siRNA Details
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130447.1 | NP_001123919.1 | 5 | 634 | ||
NM_001323630.1 | NP_001310559.1 | 5 | 594 | |||
NM_001323631.1 | NP_001310560.1 | 7 | 816 | |||
NM_001323632.1 | NP_001310561.1 | 5 | 657 | |||
NM_018097.2 | NP_060567.1 | 6 | 727 | |||
XM_017022396.1 | XP_016877885.1 | 6 | 731 | |||
XM_017022397.1 | XP_016877886.1 | 6 | 698 | |||
GenBank mRNA | AK001322.1 | - | 6 | 685 | ||
AK022939.1 | - | 6 | 698 | |||
BC010903.1 | - | 6 | 706 | 4285311 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
