siRNA Details
Gene Symbol: C14orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283057.1 | NP_001269986.1 | 5, 6 | 798 | ||
NM_001283058.1 | NP_001269987.1 | 5, 6 | 845 | |||
NM_001283059.1 | NP_001269988.1 | 5, 6 | 825 | |||
NM_018168.3 | NP_060638.2 | 5, 6 | 801 | |||
XM_005267810.3 | XP_005267867.1 | 5, 6 | 895 | |||
XM_005267811.3 | XP_005267868.1 | 4, 5 | 473 | |||
XM_005267813.1 | XP_005267870.1 | 5, 6 | 870 | |||
XM_005267814.1 | XP_005267871.1 | 5, 6 | 867 | |||
XM_006720189.2 | XP_006720252.1 | 5, 6 | 514 | |||
GenBank mRNA | AK001512.1 | - | 5, 6 | 790 | ||
AK223127.1 | - | 5, 6 | 789 | |||
BC117405.1 | - | 5, 6 | 692 | 40125956 | ||
BC143565.1 | - | 5, 6 | 739 | |||
BC143566.1 | - | 5, 6 | 692 | 9052077 | ||
BC143568.1 | - | 5, 6 | 719 |
