siRNA Details
Gene Symbol: C14orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283056.1 | NP_001269985.1 | 4 | 619 | ||
NM_001283057.1 | NP_001269986.1 | 4 | 619 | |||
NM_001283058.1 | NP_001269987.1 | 4 | 666 | |||
NM_001283059.1 | NP_001269988.1 | 4 | 646 | |||
NM_001283060.1 | NP_001269989.1 | 4 | 599 | |||
NM_018168.3 | NP_060638.2 | 4 | 622 | |||
XM_005267806.1 | XP_005267863.1 | 4 | 615 | |||
XM_005267810.3 | XP_005267867.1 | 4 | 716 | |||
XM_005267811.3 | XP_005267868.1 | 3 | 294 | |||
XM_005267813.1 | XP_005267870.1 | 4 | 691 | |||
XM_005267814.1 | XP_005267871.1 | 4 | 688 | |||
XM_006720188.2 | XP_006720251.1 | 4 | 615 | |||
XM_006720189.2 | XP_006720252.1 | 4 | 335 | |||
GenBank mRNA | AK001512.1 | - | 4 | 611 | ||
AK223127.1 | - | 4 | 610 | |||
AK307238.1 | - | 4 | 682 | |||
BC028060.1 | - | 4 | 594 | |||
BC117405.1 | - | 4 | 513 | 40125956 | ||
BC143565.1 | - | 4 | 560 | |||
BC143566.1 | - | 4 | 513 | 9052077 | ||
BC143567.1 | - | 4 | 513 | 9052078 | ||
BC143568.1 | - | 4 | 540 | |||
BI760135.1 | - | 4 | 593 |
