siRNA Details
Gene Symbol: KIAA1551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018169.3 | NP_060639.3 | 4 | 2632 | ||
XM_005253405.2 | XP_005253462.1 | 3 | 3083 | |||
XM_006719099.2 | XP_006719162.1 | 4 | 3251 | |||
XM_011520722.1 | XP_011519024.1 | 4 | 2792 | |||
XM_017019550.1 | XP_016875039.1 | 3 | 3492 | |||
GenBank mRNA | AB046771.1 | - | 1 | 1721 | ||
AK000688.1 | - | 1 | 877 | |||
AK000703.1 | - | 1 | 1473 | |||
BC140819.1 | - | 4 | 2583 | 9021687 |
