siRNA Details
Gene Symbol: PNMAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018215.3 | NP_060685.2 | 2 | 1436 | ||
XM_011527067.2 | XP_011525369.1 | 2 | 1450 | |||
GenBank mRNA | AK001643.1 | - | 1 | 455 | ||
AK096326.1 | - | 2 | 996 | |||
AK290218.1 | - | 3 | 1319 | |||
BC026026.1 | - | 1 | 469 | |||
BC032508.1 | - | 2 | 1414 | 5246377 | ||
BC051688.1 | - | 2 | 1393 | 5295669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
