siRNA Details
Gene Symbol: PNMAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103149.1 | NP_001096619.1 | 2 | 487 | ||
NM_018215.3 | NP_060685.2 | 2 | 487 | |||
XM_011527067.2 | XP_011525369.1 | 2 | 501 | |||
GenBank mRNA | AK096326.1 | - | 1 | 112 | ||
AK290218.1 | - | 2 | 435 | |||
AL834344.1 | - | 2 | 331 | |||
BC032508.1 | - | 2 | 465 | 5246377 | ||
BC051688.1 | - | 2 | 444 | 5295669 | ||
DA219062.1 | - | 2 | 462 | |||
DA325514.1 | - | 2 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
