siRNA Details
Gene Symbol: ZNF444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253792.1 | NP_001240721.1 | 5 | 2030 | ||
NM_018337.3 | NP_060807.2 | 5 | 2033 | |||
XM_005259034.2 | XP_005259091.1 | 6 | 2138 | |||
XM_005259035.1 | XP_005259092.1 | 5 | 2070 | |||
XM_005259036.1 | XP_005259093.1 | 5 | 1937 | |||
XM_011527068.1 | XP_011525370.1 | 5 | 2096 | |||
XM_011527069.1 | XP_011525371.1 | 6 | 1988 | |||
XM_011527070.1 | XP_011525372.1 | 6 | 3682 | |||
XM_011527071.1 | XP_011525373.1 | 6 | 2109 | |||
XM_011527073.1 | XP_011525375.1 | 6 | 3679 | |||
XM_017026932.1 | XP_016882421.1 | 6 | 1971 | |||
XM_017026933.1 | XP_016882422.1 | 3 | 2282 | |||
GenBank mRNA | AB052954.1 | - | 5 | 1873 | ||
AB052955.1 | - | 5 | 1876 | |||
AF074985.1 | - | 1 | 576 | |||
AK001999.1 | - | 5 | 1875 | |||
AK074063.1 | - | 2 | 5588 | |||
AK097319.1 | - | 2 | 3261 | |||
AL834130.1 | - | 5 | 2013 | |||
BC021282.2 | - | 5 | 1860 | 5016494 |
