siRNA Details
Gene Symbol: CPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018340.2 | NP_060810.2 | 3 | 438 | ||
GenBank mRNA | AK002013.1 | - | 3 | 421 | ||
AK022710.1 | - | 3 | 388 | |||
AK225359.1 | - | 3 | 388 | |||
AK225617.1 | - | 3 | 433 | |||
AY553877.1 | - | 3 | 327 | |||
BC006289.2 | - | 3 | 422 | 3959701 | ||
DA387854.1 | - | 3 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
