siRNA Details
Gene Symbol: AP5S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204446.1 | NP_001191375.1 | 3 | 1877 | ||
NM_001204447.1 | NP_001191376.1 | 3 | 1725 | |||
NM_018347.2 | NP_060817.1 | 3 | 1781 | |||
GenBank mRNA | AK002030.1 | - | 3 | 1749 | ||
AK093332.1 | - | 3 | 1693 |
