siRNA Details
Gene Symbol: TMA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018352.2 | NP_060822.2 | 7 | 847 | ||
GenBank mRNA | AK002046.1 | - | 7 | 678 | ||
BC011842.2 | - | 7 | 664 | 4110177 | ||
BX370094.1 | - | 1 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
