siRNA Details
Gene Symbol: C5orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018356.2 | NP_060826.2 | 4 | 586 | ||
XM_006714479.2 | XP_006714542.1 | 4 | 468 | |||
XM_011514062.2 | XP_011512364.1 | 5 | 664 | |||
XM_017009607.1 | XP_016865096.1 | 5 | 668 | |||
XM_017009608.1 | XP_016865097.1 | 4 | 586 | |||
XM_017009609.1 | XP_016865098.1 | 4 | 468 | |||
GenBank mRNA | AK002055.1 | - | 4 | 573 | ||
BC021215.2 | - | 4 | 523 | 4302543 |
