siRNA Details
Gene Symbol: LGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018490.2 | NP_060960.2 | 18 | 3067 | ||
GenBank mRNA | AB209743.1 | - | 18 | 2986 | ||
AF257182.1 | - | 18 | 3026 | |||
BC033039.2 | - | 17 | 2983 | 5260923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
