siRNA Details
Gene Symbol: DNAJA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130182.1 | NP_001123654.1 | 4 | 746 | ||
NM_001130183.1 | NP_001123655.1 | 4 | 573 | |||
NM_018602.3 | NP_061072.3 | 5 | 885 | |||
XM_005254535.3 | XP_005254592.1 | 5 | 658 | |||
XM_006720601.1 | XP_006720664.1 | 5 | 660 | |||
GenBank mRNA | AF116663.1 | - | 3 | 977 | ||
AK090898.1 | - | 1 | 217 | |||
AK096616.1 | - | 4 | 746 | |||
AK098079.1 | - | 5 | 876 | |||
BC021720.2 | - | 5 | 885 | 4831235 | ||
BC031044.1 | - | 4 | 608 | 5277834 | ||
BX648710.1 | - | 5 | 767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
