siRNA Details
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 56 | 6595 | ||
NM_014689.2 | NP_055504.2 | 56 | 6738 | |||
XM_005246673.4 | XP_005246730.1 | 56 | 6597 | |||
XM_006712617.3 | XP_006712680.1 | 57 | 6838 | |||
XM_017004447.1 | XP_016859936.1 | 57 | 6813 | |||
GenBank mRNA | AB074179.1 | - | 11 | 1328 | ||
AF250396.1 | - | 17 | 2090 | |||
AK000227.1 | - | 14 | 1706 | |||
AK001253.1 | - | 11 | 1338 | |||
AK293847.1 | - | 23 | 2740 | |||
AK293879.1 | - | 28 | 3127 | |||
AK307914.1 | - | 16 | 2099 | |||
BC015018.1 | - | 14 | 1705 | |||
CR749492.1 | - | 31 | 3583 | |||
EU236710.2 | - | 56 | 6592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
