siRNA Details
Gene Symbol: LRRC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017768.4 | NP_060238.3 | 6 | 898 | ||
XM_005271013.1 | XP_005271070.1 | 5 | 725 | |||
XM_011541763.1 | XP_011540065.1 | 4 | 640 | |||
XM_017001695.1 | XP_016857184.1 | 5 | 646 | |||
XM_017001696.1 | XP_016857185.1 | 3 | 420 | |||
GenBank mRNA | AK000338.1 | - | 4 | 408 | ||
AK022951.1 | - | 6 | 812 | |||
AK225337.1 | - | 4 | 408 | |||
BC003407.1 | - | 6 | 698 | |||
BC008586.1 | - | 6 | 796 | 2900193 |
