siRNA Details
Gene Symbol: SYBU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099743.1 | NP_001093213.1 | 3 | 575 | ||
NM_001099744.1 | NP_001093214.1 | 3 | 578 | |||
NM_001099745.1 | NP_001093215.1 | 3 | 414 | |||
NM_001099747.1 | NP_001093217.1 | 3 | 332 | |||
NM_001099748.1 | NP_001093218.1 | 3 | 335 | |||
NM_001099750.1 | NP_001093220.1 | 4 | 456 | |||
NM_001099751.1 | NP_001093221.1 | 3 | 421 | |||
NM_001099752.1 | NP_001093222.1 | 3 | 424 | |||
NM_001099753.1 | NP_001093223.1 | 2 | 222 | |||
NM_001099754.1 | NP_001093224.1 | 2 | 225 | |||
NM_001099756.1 | NP_001093226.1 | 1 | 296 | |||
NM_017786.5 | NP_060256.3 | 4 | 453 | |||
GenBank mRNA | AB195676.1 | - | 4 | 386 | ||
AB195677.1 | - | 3 | 265 | |||
AB195679.1 | - | 1 | 781 | |||
AK055246.1 | - | 3 | 345 | |||
AK096558.1 | - | 2 | 220 | |||
AK290469.1 | - | 3 | 414 | |||
AL713645.1 | - | 4 | 347 | |||
AY705385.1 | - | 2 | 62 | |||
BG702715.1 | - | 3 | 392 | |||
CA395128.1 | - | 4 | 313 | |||
DA155137.1 | - | 3 | 542 | |||
DA773983.1 | - | 2 | 176 | |||
DA787454.1 | - | 3 | 424 | |||
DC350594.1 | - | 3 | 414 | |||
DN993094.1 | - | 3 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
