siRNA Details
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 2 | 535 | ||
NM_001318717.1 | NP_001305646.1 | 2 | 456 | |||
NM_001318718.1 | NP_001305647.1 | 2 | 436 | |||
NM_001318719.1 | NP_001305648.1 | 2 | 414 | |||
NM_001318720.1 | NP_001305649.1 | 2 | 408 | |||
NM_001318721.1 | NP_001305650.1 | 2 | 383 | |||
NM_001318722.1 | NP_001305651.1 | 2 | 404 | |||
NM_001318723.1 | NP_001305652.1 | 2 | 488 | |||
NM_001318724.1 | NP_001305653.1 | 2 | 488 | |||
NM_001318725.1 | NP_001305654.1 | 2 | 391 | |||
NM_016506.6 | NP_057590.3 | 2 | 492 | |||
NM_018095.5 | NP_060565.4 | 2 | 387 | |||
XM_017018009.1 | XP_016873498.1 | 2 | 371 | |||
GenBank mRNA | AK001312.1 | - | 2 | 341 | ||
AK001749.1 | - | 2 | 428 | |||
AK091607.1 | - | 1 | 1065 | |||
AK222550.1 | - | 2 | 338 | |||
AK222985.1 | - | 2 | 348 | |||
AK225062.1 | - | 2 | 338 | |||
AL713735.1 | - | 1 | 317 | |||
BC002736.2 | - | 2 | 429 | 3628290 | ||
BI601113.1 | - | 2 | 395 | |||
BI908078.1 | - | 2 | 394 | |||
CR457270.1 | - | 1 | 275 | |||
DA016320.1 | - | 2 | 387 | |||
DA170281.1 | - | 2 | 443 | |||
DA559313.1 | - | 2 | 446 | |||
DA883418.1 | - | 2 | 425 | |||
DB457928.1 | - | 2 | 359 | |||
HY062454.1 | - | 2 | 370 | |||
HY120885.1 | - | 2 | 369 | |||
HY176105.1 | - | 2 | 411 |

More Information
Other Species Targeted

