siRNA Details
Gene Symbol: ASUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018164.2 | NP_060634.2 | 13 | 2042 | ||
XM_005253422.3 | XP_005253479.1 | 12 | 1830 | |||
XM_011520750.2 | XP_011519052.1 | 13 | 1593 | |||
XM_011520751.2 | XP_011519053.1 | 12 | 1848 | |||
XM_011520752.2 | XP_011519054.1 | 12 | 1794 | |||
XM_011520753.2 | XP_011519055.1 | 7 | 765 | |||
XM_017019632.1 | XP_016875121.1 | 13 | 1575 | |||
XM_017019633.1 | XP_016875122.1 | 12 | 1985 | |||
XM_017019634.1 | XP_016875123.1 | 12 | 1967 | |||
XM_017019635.1 | XP_016875124.1 | 12 | 1404 | |||
XM_017019636.1 | XP_016875125.1 | 12 | 1776 | |||
XM_017019637.1 | XP_016875126.1 | 11 | 1205 | |||
GenBank mRNA | AF274950.1 | - | 4 | 454 | ||
AK001222.1 | - | 3 | 357 | |||
AK001492.1 | - | 13 | 1671 | |||
AK001499.1 | - | 13 | 1707 | |||
AK225336.1 | - | 13 | 1693 | |||
AK297948.1 | - | 11 | 1382 | |||
AY211927.1 | - | 6 | 704 | |||
BC003081.2 | - | 13 | 1731 | 2989678 | ||
BC008368.1 | - | 13 | 1689 | 3355282 |

More Information
Other Species Targeted

Gene Ontology Categories:
