siRNA Details
Gene Symbol: DNAJC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018198.3 | NP_060668.2 | 11 | 1323 | ||
XM_017001745.1 | XP_016857234.1 | 10 | 1176 | |||
GenBank mRNA | AF306695.1 | - | 11 | 1237 | ||
AK001599.1 | - | 11 | 1232 | |||
AK095386.1 | - | 3 | 254 | |||
AK294539.1 | - | 10 | 1146 | |||
AK298378.1 | - | 11 | 1209 | |||
AL833841.1 | - | 11 | 1197 | |||
AY336749.1 | - | 11 | 1233 | |||
BC006086.1 | - | 11 | 1219 | 3946414 | ||
BC008772.1 | - | 11 | 1224 |
