siRNA Details
Gene Symbol: C1orf106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142569.2 | NP_001136041.1 | 4 | 620 | ||
NM_018265.3 | NP_060735.3 | 4 | 556 | |||
XM_011509754.2 | XP_011508056.1 | 4 | 1335 | |||
XM_011509755.1 | XP_011508057.1 | 4 | 597 | |||
XM_011509756.2 | XP_011508058.1 | 4 | 594 | |||
GenBank mRNA | AK001763.1 | - | 4 | 514 | ||
AK303885.1 | - | 4 | 521 | |||
AK316510.1 | - | 4 | 620 | |||
BC106877.2 | - | 4 | 514 | 40026425 | ||
BP357953.1 | - | 4 | 555 |
