siRNA Details
Gene Symbol: ZNF839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267827.1 | NP_001254756.1 | 6 | 1673 | ||
NM_001267828.1 | NP_001254757.1 | 6 | 1604 | |||
NM_018335.4 | NP_060805.3 | 6 | 1686 | |||
XM_006720203.3 | XP_006720266.2 | 6 | 1570 | |||
XM_011536945.1 | XP_011535247.1 | 7 | 1788 | |||
XM_011536946.2 | XP_011535248.1 | 6 | 1638 | |||
XM_011536948.2 | XP_011535250.1 | 6 | 1414 | |||
XM_011536949.2 | XP_011535251.1 | 7 | 1788 | |||
XM_011536950.2 | XP_011535252.1 | 6 | 1638 | |||
XM_017021447.1 | XP_016876936.1 | 5 | 1536 | |||
XM_017021448.1 | XP_016876937.1 | 6 | 1686 | |||
XM_017021449.1 | XP_016876938.1 | 5 | 952 | |||
XM_017021450.1 | XP_016876939.1 | 7 | 3869 | |||
XM_017021451.1 | XP_016876940.1 | 6 | 1592 | |||
GenBank mRNA | AF155112.1 | - | 4 | 315 | ||
AK001994.1 | - | 2 | 102 | |||
AK093342.1 | - | 6 | 1673 | |||
AK125678.1 | - | 2 | 822 | |||
AK127821.1 | - | 2 | 920 | |||
AK223312.1 | - | 2 | 86 | |||
AK289632.1 | - | 6 | 1604 | |||
AL137478.1 | - | 5 | 960 | |||
BC073137.1 | - | 4 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
