siRNA Details
Gene Symbol: ZNF415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136038.2 | NP_001129510.2 | 3, 4 | 507 | ||
NM_001164309.1 | NP_001157781.1 | 3, 4 | 380 | |||
NM_018355.3 | NP_060825.2 | 3, 4 | 275 | |||
XM_006723268.1 | XP_006723331.1 | 3, 4 | 341 | |||
XM_011527103.1 | XP_011525405.1 | 4, 5 | 412 | |||
XM_017026964.1 | XP_016882453.1 | 4, 5 | 495 | |||
XM_017026965.1 | XP_016882454.1 | 3, 4 | 509 | |||
XM_017026966.1 | XP_016882455.1 | 4, 5 | 637 | |||
XM_017026967.1 | XP_016882456.1 | 3, 4 | 435 | |||
GenBank mRNA | AK002053.1 | - | 3, 4 | 247 | ||
AY283600.1 | - | 3, 4 | 247 | |||
DA075573.1 | - | 3, 4 | 362 | |||
DA518713.1 | - | 3, 4 | 477 | |||
DB285275.1 | - | 3, 4 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
