siRNA Details
Gene Symbol: ADCY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167749.2 | NP_001161221.1 | 28 | 4247 | ||
NM_001297772.1 | NP_001284701.1 | 31 | 4739 | |||
NM_018417.5 | NP_060887.2 | 31 | 4702 | |||
XM_006711449.3 | XP_006711512.1 | 31 | 6032 | |||
XM_011509760.2 | XP_011508062.1 | 32 | 6172 | |||
XM_011509762.2 | XP_011508064.1 | 30 | 5888 | |||
XM_011509763.2 | XP_011508065.1 | 30 | 5870 | |||
GenBank mRNA | AF176813.2 | - | 31 | 4578 | ||
AF271058.1 | - | 30 | 4393 | |||
AF299350.1 | - | 31 | 4578 | |||
AF331033.1 | - | 31 | 4615 | |||
AK302884.1 | - | 28 | 4236 | |||
AL035122.1 | - | 13 | 2095 | |||
BC117366.1 | - | 31 | 4482 | 40125917 | ||
BC117372.1 | - | 31 | 4482 | 40125923 | ||
GQ472219.1 | - | 31 | 4482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
