siRNA Details
Gene Symbol: ADCY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167749.2 | NP_001161221.1 | 15 | 2091 | ||
NM_001297772.1 | NP_001284701.1 | 18 | 2583 | |||
NM_018417.5 | NP_060887.2 | 18 | 2546 | |||
XM_005245330.4 | XP_005245387.1 | 18 | 3876 | |||
XM_006711449.3 | XP_006711512.1 | 18 | 3876 | |||
XM_011509760.2 | XP_011508062.1 | 19 | 4016 | |||
XM_011509762.2 | XP_011508064.1 | 17 | 3732 | |||
XM_011509763.2 | XP_011508065.1 | 18 | 3876 | |||
XM_011509766.2 | XP_011508068.1 | 18 | 3876 | |||
XM_017001778.1 | XP_016857267.1 | 18 | 3876 | |||
GenBank mRNA | AF176813.2 | - | 18 | 2422 | ||
AF271058.1 | - | 17 | 2237 | |||
AF299350.1 | - | 18 | 2422 | |||
AF331033.1 | - | 18 | 2459 | |||
AK302884.1 | - | 15 | 2080 | |||
AK310395.1 | - | 18 | 2433 | |||
BC117366.1 | - | 18 | 2326 | 40125917 | ||
BC117372.1 | - | 18 | 2326 | 40125923 | ||
GQ472219.1 | - | 18 | 2326 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
