siRNA Details
Gene Symbol: CENPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018455.5 | NP_060925.2 | 7 | 1360 | ||
XM_017023456.1 | XP_016878945.1 | 7 | 760 | |||
GenBank mRNA | AF217515.1 | - | 7 | 910 | ||
BC007334.2 | - | 7 | 1306 | 3830110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
