siRNA Details
Gene Symbol: WWC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015691.3 | NP_056506.2 | 20, 21 | 3047 | ||
GenBank mRNA | AB033106.1 | - | 18, 19 | 2670 | ||
AK024683.1 | - | 11, 12 | 1857 | |||
AK091936.1 | - | 11, 12 | 1724 | |||
AK292961.1 | - | 19, 20 | 2929 | |||
AL353937.1 | - | 17, 18 | 2538 | |||
BC003527.1 | - | 2, 3 | 224 | |||
BC032447.1 | - | 17, 18 | 2582 | |||
KC987947.1 | - | 20, 21 | 3224 |
