siRNA Details
Gene Symbol: ZC3H15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018471.2 | NP_060941.2 | 4 | 579 | ||
XM_017004502.1 | XP_016859991.1 | 4 | 484 | |||
GenBank mRNA | AF109366.1 | - | 4 | 438 | ||
AF161421.1 | - | 4 | 450 | |||
AF220184.1 | - | 4 | 580 | |||
AF289586.1 | - | 4 | 479 | |||
AK001901.1 | - | 4 | 484 | |||
AK298615.1 | - | 533 | ||||
BC021102.1 | - | 4 | 391 | 5020890 |
