siRNA Details
Gene Symbol: ECHDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002030.1 | NP_001002030.1 | 2 | 327 | ||
NM_001139510.1 | NP_001132982.1 | 2 | 105 | |||
NM_018479.3 | NP_060949.2 | 2 | 327 | |||
XM_005267047.2 | XP_005267104.1 | 3 | 252 | |||
XM_005267048.1 | XP_005267105.1 | 2 | 426 | |||
XM_005267049.2 | XP_005267106.1 | 2 | 594 | |||
XM_005267050.2 | XP_005267107.1 | 3 | 220 | |||
XM_011535942.2 | XP_011534244.1 | 2 | 182 | |||
XM_011535943.2 | XP_011534245.1 | 2 | 597 | |||
XM_017011056.1 | XP_016866545.1 | 2 | 182 | |||
GenBank mRNA | AF220192.1 | - | 2 | 323 | ||
AK025760.1 | - | 2 | 77 | |||
AK025796.1 | - | 2 | 105 | |||
AL834469.1 | - | 2 | 266 | |||
BC003549.1 | - | 2 | 289 | 3547005 | ||
BQ224769.1 | - | 2 | 273 | |||
DC297239.1 | - | 2 | 327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
