siRNA Details
Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 6 | 1506 | ||
GenBank mRNA | AB035966.1 | - | 6 | 983 | ||
AF353942.1 | - | 6 | 983 | |||
AJ278245.1 | - | 6 | 1114 | |||
AK095857.1 | - | 6 | 1506 | |||
AK225958.1 | - | 6 | 959 | |||
AK301747.1 | - | 6 | 912 | |||
AK313326.1 | - | 6 | 1251 | |||
AL713665.2 | - | 6 | 898 | |||
BC004887.2 | - | 3 | 334 | |||
BC070049.1 | - | 6 | 1202 | 30343188 |
