siRNA Details
Gene Symbol: NXT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242617.1 | NP_001229546.1 | 4 | 473 | ||
NM_001242618.1 | NP_001229547.1 | 4 | 434 | |||
NM_018698.4 | NP_061168.2 | 5 | 563 | |||
XM_011530989.2 | XP_011529291.1 | 4 | 476 | |||
XM_017029648.1 | XP_016885137.1 | 4 | 411 | |||
GenBank mRNA | AF201942.1 | - | 4 | 440 | ||
AF212223.1 | - | 5 | 425 | |||
AF246127.1 | - | 3 | 296 | |||
AJ277591.1 | - | 3 | 296 | |||
AJ278323.1 | - | 4 | 434 | |||
AK023289.1 | - | 5 | 563 | |||
BC014888.1 | - | 4 | 427 | |||
BC120984.1 | - | 5 | 552 | 40116082 | ||
BC120985.1 | - | 5 | 552 | 40116088 | ||
DA442845.1 | - | 4 | 475 |
