siRNA Details
Gene Symbol: PCDHB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018937.4 | NP_061760.1 | 1 | 566 | ||
GenBank mRNA | AF152496.1 | - | 1 | 158 | ||
AF217755.1 | - | 1 | 158 | |||
AK313451.1 | - | 1 | 335 | |||
BC104817.1 | - | 1 | 185 | 8143820 |
