siRNA Details
Gene Symbol: MTFR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099625.1 | NP_001093095.1 | 6 | 959 | ||
NM_001099626.1 | NP_001093096.1 | 6 | 1011 | |||
NM_001099627.1 | NP_001093097.1 | 6 | 850 | |||
NM_019557.5 | NP_062457.3 | 6 | 1057 | |||
GenBank mRNA | AB075880.1 | - | 6 | 887 | ||
AF173891.1 | - | 6 | 824 | |||
AK056721.1 | - | 5 | 2213 | |||
AK127164.1 | - | 2 | 1493 | |||
AK290744.1 | - | 5 | 646 | |||
AK299221.1 | - | 6 | 902 | |||
AK314297.1 | - | 6 | 887 | |||
AL512766.1 | - | 6 | 1059 | |||
AY007100.1 | - | 6 | 937 | |||
BC017175.2 | - | 6 | 1011 | 3162883 | ||
BX647921.1 | - | 6 | 807 | |||
KU178723.1 | - | 5 | 749 | |||
KU178724.1 | - | 5 | 772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
